missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Viral Nucleic Acid Extraction Kit

For extraction of Viral DNA or RNA from human biological specimens such as serum, plasma, CSF, urine and other cell-free fluids

Brand:  MP Biomedicals™ 117022300


Product Code. 15739210

  • £324.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Completely automated purification protocol
  • Run in as little as 40 minutes
  • Nucleic Acids can be used immediately in downstream applications such as PCR, qPCR, Sequencing (NGS), Microarray, RFLP, and Southern Blot Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature
Magnetic Bead
MPure-12 Automated Nucleic Acid Purification System
Proteinase K solution, 30μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL: RNase-free water, 1000μL; RNase-free water, 1000μL
100 to 400μL
Viral Nucleic Acid Extraction Kit
Human Biological Specimens (Serum, Plasma, and Other Cell-free Fluids)
Viral DNA or RNA Purification
48 Preps
As little as 40 minutes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MPure Viral Nucleic Acid Extraction Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glycerol, proteinaseK, guanidine thiocyanate, triton X-100,isopropyl alcohol,ethyl alcohol, guani

SDS
Documents

Documents

Certificates
Special Offers

Special Offers