missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MPure™ Viral Nucleic Acid Extraction Kit 800
For extraction of Viral DNA or RNA from human biological specimens such as serum, plasma, and other cell-free fluids
Brand: MP Biomedicals™ 117022170
This item is not returnable.
View return policy
Description
- Used with the MPure-12 instrument for extraction
- Completely automated purification protocol
- Run in as little as 55 minutes
- Nucleic acids can be used immediately in downstream applications
Specifications
Store at room temperature | |
Human Biological Specimens (Serum, Plasma, and Other Cell-free Fluids) | |
Nucleic Acid Purification; Viral DNA or RNA from human biological specimens such as serum, plasma, and other cell-free fluids | |
48 Preps |
Viral Nucleic Acid Extraction Kit 800 | |
MPure-12 Automated Nucleic Acid Purification System | |
Proteinase K Solution: 40μL; Lysis Buffer 2: 1000μL; Binding Buffer 1: 1600μL; Magnetic Bead Solution: 800μL; Washing Buffer 1: 1000μL; Washing Buffer 2: 1000μL; Washing Buffer 3: 1000μL; RNase Free Water: 1000μL | |
600 to 800μL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MPure Viral Nucleic Acid Extraction Kit 800
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Proteinase K