missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Viral/Pathogen Nucleic Acids Extraction Kit A

MPure Viral/Pathogen Nucleic Acids Extraction Kit A extracts viral and bacterial DNA/RNA from cell-free samples including serum, plasma and other cell-free body fluids.

Brand:  MP Biomedicals™ 117022120


Product Code. 15779200

  • £327.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Run in as little as 40 minutes
  • Nucleic acid is ready for immediate use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature
MPure-12 Automated Nucleic Acid Purification System
Reagent Cartridge: 48, Reaction Chamber: 48, Tip Holder: 48, Filtered Tip: 50, Piercing Pin: 50, Sample Tube (2 mL): 50, Elute Tube (1.5 mL): 50, RNA Carrier (1 mg): 1, Barcode Paper: 1, Proteinase K solution: 40 μL, Lysis Buffer 1: 720 μL, Binding Buffer 1: 1000 μL, Magnetic Bead Solution: 800 μL, Washing Buffer 1: 1000 μL, Washing Buffer 2: 1000 μL, Washing Buffer 3: 1000 μL, RNase-free water: 1000 μL, BL2 Buffer: 400 μL
Viral/Pathogen Nucleic Acid Extraction Kit
As little as 40 minutes
Kit
Nucleic Acid Purification, DNA and RNA
48 Preps
100 to 400 μL (virus), 100 to 200 μL (virus/bacteria)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MPure Viral/Pathogen Nucleic Acids Extraction Kit A

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Proteinase K

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.