Learn More
Tocris Bioscience™ MQAE
Fluorescent Cl- indicator; membrane permeable
Brand:  Tocris Bioscience™ 7856/100
Product Code. 18766335
  • £103.00 / 100mg
Explore more special offers
Description
Description
MQAE is a membrane-permeable intracellular chloride ion (Cl-) indicator. The fluorescence intensity of MQAE decreases proportionally as Cl- ions increase. Excitation/emission maxima (λ) are 355 nm and 460 nm respectively. Can be used in fluorescence microscopy and flow cytometry, suitable for use in vivo and in vitro.
Specifications
Specifications
1-(2-Ethoxy-2-oxoethyl)-6-methoxyquinolin-1-ium bromide
100 mg
C14H16BrNO3
162558-52-3
Ion Indicators
0.95