missing translation for 'onlineSavingsMsg'
Learn More

MRS 2179 tetrasodium salt, Tocris Bioscience™

Product Code. 18748576
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective P2Y1 antagonist

A competitive antagonist at P2Y1 receptors (KB = 100 nM). Selective over P2X1 (IC50 = 1.15 μM), P2X3 (IC50 = 12.9 μM), P2X2, P2X4, P2Y2, P2Y4 and P2Y6 receptors. Inhibits the upregulation of NTPDase1 by ATPγS.

Chemical Identifiers

CAS 1454889-37-2
Molecular Formula C11H17N5Na4O9P2
Molecular Weight (g/mol) 517.19
InChI Key HLJPLHFSEAJJGC-ZKRIHRHSSA-N
Synonym 2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
PubChem CID 131848299
IUPAC Name [(2R,3S,5R)-5-[6-(methylamino)purin-9-yl]-2-(phosphonooxymethyl)oxolan-3-yl] dihydrogen phosphate;sodium
SMILES CNC1=NC=NC2=C1N=CN2C3CC(C(O3)COP(=O)(O)O)OP(=O)(O)O.[Na].[Na].[Na].[Na]

Specifications

Quantity 10 mg
Formula Weight 513.16
Percent Purity >98%
Chemical Name or Material MRS 2179 tetrasodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'