Learn More
Tocris Bioscience™ MRS 2578
Selective P2Y6 antagonist
£132.00 - £475.00
Specifications
Product Type MRS 2578
Molecular Weight (g/mol) 472.66
Inhibitors MRS 2578
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18745586
View Documents Promotion Details
Tocris Bioscience™
2146/10
10 mg
£132.00
10mg
18004492
View Documents
Tocris Bioscience™
2146/50
50 mg
£475.00
50mg
Description
Description
Selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
Specifications
Specifications
MRS 2578
472.66
MRS 2578