missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRS 2768 tetrasodium salt, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Selective P2Y2 agonist (EC50 = 1.89 μM). Displays no affinity for human P2Y4 or P2Y6 receptors.
Specifications
Specifications
CAS | 2567869-47-8 |
Molecular Formula | C15H16N2Na4O18P4 |
Quantity | 1 mg |
Synonym | mrs 2768 tetrasodium salt, uridine-5'-tetraphosphate ?-phenyl ester tetrasodium salt |
InChI Key | ASYFBNZFGLWLNC-YYXHNCPRSA-J |
SMILES | C1=CC=C(C=C1)OP(=O)([O-])OP(=O)([O-])OP(=O)([O-])OP(=O)([O-])OCC2C(C(C(O2)N3C=CC(=O)NC3=O)O)O.[Na+].[Na+].[Na+].[Na+] |
IUPAC Name | tetrasodium;[[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [oxido-[oxido(phenoxy)phosphoryl]oxyphosphoryl] phosphate |
Molecular Weight (g/mol) | 728.143 |
PubChem CID | 90488893 |
Formula Weight | 728.14 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction