missing translation for 'onlineSavingsMsg'
Learn More

MRS 4062 triethylammonium salt, Tocris Bioscience™

Product Code. 18004232
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective P2Y4 receptor agonist

Selective P2Y4 receptor agonist (EC50 values are 23, 640, and 740 nM for hP2Y4, hP2Y2 and hP2Y6 respectively).

Specifications

CAS 1309871-50-8
Molecular Formula C42H86N7O15P3
Quantity 1 mg
Synonym mrs 4062 triethylammonium salt, n4-phenylpropoxycytidine-5'-o-triphosphate tetra triethylammonium salt
InChI Key ZCDZFMKRHPANNI-IHQKQWLFSA-N
SMILES CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC.C1=CC=C(C=C1)CCCONC2=NC(=O)N(C=C2)C3C(C(C(O3)COP(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-])O)O
IUPAC Name [[[(2R,3S,4R,5R)-3,4-dihydroxy-5-[2-oxo-4-(3-phenylpropoxyamino)pyrimidin-1-yl]oxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl] phosphate;triethylazanium
Molecular Weight (g/mol) 1022.105
PubChem CID 90488925
Formula Weight 1022.09
Percent Purity 98%
Chemical Name or Material MRS 4062 triethylammonium salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'