Learn More
Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets
Conserve energy with these cabinets, which combine smart design and extraordinary value with best-in-class energy efficiency, reliability and usability.
£8805.37 - £11449.65
Specifications
Certifications/Compliance | EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) |
---|---|
Product Type | Biosafety Cabinet |
Exhaust Air Filter | HEPA H 14 EN 1822, 99.995% at 0.3µm Particle |
Green Features | Energy efficient |
Ports | TUV-approved |
Product Code | Brand | Width (English) Interior | Width (Metric) Interior | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (English) Interior | Width (Metric) Interior | Price | Quantity & Availability | ||||
10795194
|
Thermo Scientific™
51025411 |
4 ft. | 1.2 m |
£9512.69
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10445753
|
Thermo Scientific™
51025413 |
6 ft. | 1.8 m |
£11449.65
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11762997
|
Thermo Scientific™
51028225 |
2.95 ft. | 0.9 m |
£8805.37
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11752997
|
Thermo Scientific™
51028226 |
4.92 ft. | 1.5 m |
£11103.73
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets (BSCs) combine safety and extraordinary value with energy efficiency, reliability and usability. With proprietary airflow design and energy-efficient DC motors, the MSC-Advantage BSCs use up to 68% less energy than cabinets with traditional AC motors. The cabinets provide both personal and product protection from biological hazards and contamination while affording users with a comfortable and ergonomic workspace. Cabinets are certified to EN 12469 safety standard.
The MSC-Advantage biological safety is available in 4 sizes and can be connected to an external exhaust to provide protection for trace amounts of volatile toxic chemicals that are sometimes used in the cabinet.
- Available in 90 cm, 120 cm, 150 cm and 180 cm
Containment
- Thermo Scientific™ SmartFlow™ technology automates airflow balancing and compensation
- Digital Airflow Verification (DAVe) alerts user to compromised product and operator protection
- H14 HEPA filters remove airborne contaminants
- SmartFlow™ indicator displays performance status
Comfort
- Low noise level enhances user focus
- 10° sloped front for exceptional comfort and ergonomics
- Spacious work area with comfortable armrests
- Increased light intensity for a brighter work environment
Convenience
- SmartClean front window design to improve cleaning and loading of large equipment
- Intuitive control panel displays safety and performance data
- Thermo Scientific™ SmartPort™ design organises tubing and cables
- Programmable UV light extends bulb life and saves energy (UV light is optional)
Certifications/Compliance
- Fully compliant with the EN 12469 safety standard as independently tested and certified by TÜV Nord
Ordering Information:
Please order separately: Stand options, programmable UV light, one-piece work tray, ergonomic stool, free-standing floor shelf. Not available for sale in North America (equivalent model: 1300 Series A2 Biological Safety Cabinets).
Specifications
EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) | |
HEPA H 14 EN 1822, 99.995% at 0.3µm Particle | |
TUV-approved | |
80 cm |
Biosafety Cabinet | |
Energy efficient | |
152 cm | |
230 V |