missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ MSL 8 High Volume Magnetic Stirrer

Magnetic stirrer with continuous operations at both low and high speeds to ensure safety and resistance to chemical agents. MSL 8 High Volume Magnetic Stirrer is an ideal solution for large capacity laboratory stirring applications in life science and pharmaceuticals.

Brand:  Velp Scientifica™ F203A0513

Product Code. 17220464

  • £225.00 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Premium performance for large capacity laboratory stirring applications

The MSL 8 is the high volume magnetic stirrer without heating designed to guarantee the best performance for large capacity and low-viscosity laboratory stirring applications.

  • High volumes preparations
  • Buffer solution preparations
  • Buffer preparation and WFI (Water for Injection) especially in the Downstream department in Pharma
  • Sample preparation for buffer solution to be used for Dissolution in Dissolution Apparatus
  • Vaccine manufacturing
  • Cell cultures and tissue cultures (with spinner flasks)
  • Applications with nalgene carboys, as well as large volume glass bottles, are supported
Safe and Reliable:
  • The brushless motor provides continuous operation for high volumes at any speed
  • Stirs up to 8 liter volume (H2O)
  • Speed range from 40 to 1500 rpm
  • SpeedServo™ technology maintains constant speed
  • Gentle start to protect your sample
  • The control panel is protected from liquid spills
  • Resistant stainless steel cover
  • Designed for the longest lifespan
Specifications

Specifications

40 to 1500 rpm
Stainless Steel
Analog
100/240 V
Magnetic Stirrer
1
8 L
Square
4.5 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.