missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent

Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.

£110.55 - £608.03

Specifications

For Use With GC/MS
Hazards Flammable, Irritant
View More Specs

Products 5
Product Code Brand Standard Components Quantity Price Quantity & Availability  
Product Code Brand Standard Components Quantity Price Quantity & Availability  
11587670
View Documents
Thermo Scientific™
TS-48911
MSTFA (N-Methyl-N-trimethylsilyltrifluoroacetamide) 10 g
£110.55
Each
Please to purchase this item. Need a web account? Register with us today!
11786957
View Documents Promotion Details
Thermo Scientific™
TS-48910
MSTFA (N-Methyl-N-trimethylsilyltrifluoroacetamide) 10 x 1 mL ampules
£140.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11507680
View Documents Promotion Details
Thermo Scientific™
TS-48914
MSTFA (N-Methyl-N-trimethylsilyltrifluoroacetamide) 100 mL
£608.03
Each
Please to purchase this item. Need a web account? Register with us today!
11567851
View Documents Promotion Details
Thermo Scientific™
TS-48915
MSTFA plus 1% TMCS (2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)-acetamide, Chlorotrimethylsilane) 10 x 1 mL ampules
£194.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11597670
View Documents Promotion Details
Thermo Scientific™
TS-48913
MSTFA (N-Methyl-N-trimethylsilyltrifluoroacetamide) 25 mL
£281.00
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.

  • Trimethylsilyl donor strength comparable to BSA and BSTFA
  • Reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH3)3 group
  • Used to prepare volatile and thermally stable derivatives for GC and MS
  • Volatile byproduct, N-methyltrifluoroacetamide, has an even lower retention time than MSTFA
  • Often TMS derivatives of small molecules can be analyzed when derivatized with MSTFA because the byproducts and the reagent itself usually elute with the solvent front
  • Addition of Thermo Scientfic TMCS aids derivatization of amides, secondary amines and hindered hydroxyls not derivatized by MSTFA alone
Specifications

Specifications

GC/MS
Flammable, Irritant
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.