missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTCO1 Monoclonal Antibody (1D6E1A8)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ 459600

378.00 GBP valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.


Product Code. 10444035

  • £504.00 / 100µg
Seek Out Great Savings With the Lab of Curiosity
Use promo code 24110
View all promotions

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

For best results with this antibody in Western blot, do not boil samples before loading onto the gel. Boiling of the sample will cause a loss of signal. Hydrophobic intrinsic membrane proteins such as the core mtDNA-encoded proteins of the mitochondrial OXPHOS complexes tend to run faster in SDS-PAGE than predicted by their amino acid composition. This is likely due to incomplete unfolding of the protein and a more negative charge:mass ratio. In Western blot applications, this antibody detects a band at approximately 40 kDa (predicted MW: 57 kDa). In mouse liver lysate a specific band below 37 kDa was detected. Recommended positive controls: ICC/IF - Pig retinal pigment epithelial cells, rat cerebellum primary cells IHC (P) - Human skeletal muscle and colon tissue, rat pancreas tissue, pig smooth muscle tissue WB - Mouse, human, bovine and rat heart mitochondria lysate Flow - HEK-293 cells Recommended long term storage at -20°C.

Cytochrome c oxidase subunit I (CO1 or MTCO1) is 1 of 3 mitochondrial DNA (mtDNA) encoded subunits (MTCO1, MTCO2, MTCO3) of respiratory Complex IV. Complex IV is located within the mitochondrial inner membrane and is the third and final enzyme of the electron transport chain of mitochondrial oxidative phosphorylation. It is composed of 13 polypeptides. Subunits I, II, and III (MTCO1, MTCO2, MTCO3) are encoded by mtDNA while subunits IV, Va, Vb, VIa, VIb, VIc, VIIa, VIIb, VIIc, and VIII are nuclear encoded. The cytochrome c oxidase family of enzymes have 4 redox centers, 2 hemes and 2 copper centers. In mitochondrial Complex IV, the 2 hemes are a and a3 and the 2 coppers are CuA and CuB. The 2 hemes and CuB are bound to subunit I. Acin-Perez et al. (2003) identified a cell line containing single and double missense mutations in the cytochrome c oxidase (COX) subunit I gene of mouse mitochondrial DNA. And they hypothesized that deleterious mutations can arise and become predominant; cultured cells can maintain several mtDNA haplotypes at stable frequencies; the respiratory chain has little spare COX capacity; and that the size of a cavity in the vicinity of val421 in MTCO1I of animal COX may affect the function of the enzyme.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTCO1
Monoclonal
0.99 mg/mL
PBS with 0.05% BSA, 40% glycerol and 0.01% sodium azide
P00395, P00397, P05503
mt-Co1
Human Mitochondrial Complex IV subunit I.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2a κ
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1D6E1A8
Unconjugated
mt-Co1
Co1; coi; cox1; coxi; Cytochrome c oxidase polypeptide I; Cytochrome c oxidase subunit 1; cytochrome c oxidase subunit I; mitochondrially encoded cytochrome c oxidase I; mtco1; mt-co1
Mouse
Protein A
RUO
17708, 26195, 4512
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.