Learn More
STEMCELL Technologies mTeSR™3D
Defined, serum-free media for suspension culture of human ES and iPS cells
Brand: STEMCELL Technologies 03950
Additional Details : Weight : 0.50000kg
Description
- mTeSR™3D was developed for expansion and scale-up of undifferentiated human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture
- The novel fed-batch workflow saves time and media
- Daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days
- mTeSR™3D is a specialised and serum-free formulation
- This culture system is compatible with a range of suspension culture vessels
Specifications
Pluripotent Stem Cells | |
Cell Culture, Expansion | |
1 Kit |