Learn More
STEMCELL Technologies mTeSR™ Plus
cGMP, stabilized feeder-free maintenance medium for human ES and iPS cells
Brand:  STEMCELL Technologies 100-0276
Additional Details : Weight : 0.68220kg
Product Code. 17187501
  • £465.00 / 1 set
Estimated Shipment: 24-07-2024
to see stock.
Description
Description
  • mTeSR™ Plus is a serum-free, stabilized cell culture medium for the feeder-free maintenance and expansion of human embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • It is based on the mTeSR™1 formulation, the most widely published feeder-free cell culture medium for human ES and iPS cells
  • To enhance cell-quality attributes during restricted feeds, critical medium components have been stabilized, including FGF2, and medium pH is more consistent
  • As a result, mTeSR™ Plus allows for greater cell expansion rates with daily feeding, while also maintaining cell quality during restricted feeding schedules
  • mTeSR™ Plus is compatible with a variety of culture matrices, including Corning™ Matrigel™ hESC-Qualified Matrix (Corning Catalog #354277) and Vitronectin XF™ (Catalog #07180, a matrix developed and manufactured by Nucleus Biologics)
  • Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and then performance-tested in a culture assay using human pluripotent stem cells (hPSCs)
  • mTeSR™ Plus (Catalog #100-0276) is manufactured under relevant cGMPs, ensuring the highest quality and consistency for reproducible results
Specifications
Specifications
Pluripotent Stem Cells
Cell Culture, Expansion, Maintenance
1 Kit