missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552725
This item is not returnable.
View return policy
Description
Immunogen sequence: LDHLYSCRFG TFLFNCESAR ERQKVTERTV SLWSLINSNK EKFKNPFYTK EINRVLYPVA SMRHLELWVN YYIRWNPRIK QQQPNPVEQR YMELLALRDE YIKRLEELQL ANSAKLSDPP TSPSSPSQMM P Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 55%.
Human MTM1, a 603 amino-acid protein, is mutated in myotubular myopathy. The largely related protein hMTMR2 is found mutated in a recessive form of Charcot-Marie-tooth neuropathy. Myotubularin is primarily a lipid phosphatase that acts on hosphatidylinositol 3-monophosphate and is involved in the regulation of the phosphatidylinositol 3-kinase (PI 3-kinase) pathway and membrane trafficking. Wildtype myotubularin can directly dephosphorylate PI 3-P and PI 4-P in vitro. Thus, it decreases PI 3-P levels by down-regulating PI 3-K activity and by facilitating the degradation of PI 3-P.Specifications
MTM1 | |
Polyclonal | |
Unconjugated | |
MTM1 | |
AF073996; CG2; CNM; fb19c01; hypothetical protein LOC560881; im:6903724; mKIAA4176; Mtm; MTM1; MTMX; myotubularin; myotubularin 1; Myotubularin-like protein; phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; wu:fb19c01; X-linked myotubular myopathy gene 1; XLMTM; zgc:123266 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13496 | |
MTM1 | |
Recombinant protein corresponding to Human MTM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only