missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ MTT (3-(4,5-Dimethylthiazol-2-yl)-2,5-Diphenyltetrazolium Bromide)
MTT (3-(4,5-Dimethylthiazol-2-yl)-2,5-Diphenyltetrazolium Bromide)
Brand: Molecular Probes™ M6494
93.72 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
MTT is used to assess cell viability as a function of redox potential. Actively respiring cells convert the water-soluble MTT to an insoluble purple formazan. The formazan is then solubilized and its concentration determined by optical density.Specifications
Store at room temperature and protect from light. | |
Absorbance | |
Colorimetric Reagent-Based | |
1 g |
MTT (3-(4,5-Dimethylthiazol-2-yl)-2,5-Diphenyltetrazolium Bromide) | |
MTT | |
Pierce | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.