Learn More
Thermo Scientific™ Mueller Hinton Agar
Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.
£11.05 - £26.99
Product Code | Brand | Format | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Quantity | Price | Quantity & Availability | ||||
12902588
|
Thermo Scientific™
PO0152A |
90mm Monoplate | 10/Pk. |
£25.13
Pack of 10 |
In Stock
Log in to see stock. |
||||
15833237
|
Thermo Scientific™
PO1191S |
Plate | 10/Pk. |
£26.99
Pack of 5 |
In Stock
Log in to see stock. |
||||
15852467
|
Thermo Scientific™
PO5403I |
120mm Squared Plate | 4/Pk. |
£11.05
Pack of 4 |
In Stock
Log in to see stock. |
||||
Description
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.
The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.Our products are manufactured and distributed across the world through a vast network of company owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Agar |