missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES

Perform susceptibility testing with a full range of supplies, including this broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.

Brand:  Thermo Scientific™ YT3462

481.32 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15466584

  • £731.00 / Pack of 100
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform high-quality susceptibility testing with a complete range of supplies, including Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES, to complement and enhance your Thermo Scientific™ Sensititre™ test program.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES
Microbial Susceptibility
11 mL/Bottle
Microbiology Media
100
100 x 11 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.