Learn More
Technical Service Consultants™ Mulitpurpose Sampling Container
500 mL sterile multipurpose sampling container.
Brand: Technical Service Consultants™ TS/15-F40
Additional Details : Weight : 0.90000kg
Description
- Batch and expiry details are stated on the side of the label
- Recommended for physical or chemical testing
- Ideal for water sampling
- Shelf Life: 2 years
- Sterilisation Method: Gamma Irradiation to achieve a SAL of 10-6
Specifications
HDPE | |
Clear Bottle, Blue Cap | |
40 |
500 mL | |
Physical or chemical testing; water sampling |