missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin Broth (MkTTn)
Selectively isolate Salmonella species from food samples with Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin Broth.
Brand: Thermo Scientific™ CM1048B
72.74 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Selectively isolate Salmonella species from food samples with Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin Broth (MkTTn) for food testing in compliance with ISO 6579:2002.
Muller-Kauffmann Tetrathionate-Novobiocin Broth was developed by Muller1 and later modified by Kauffmann234 with the addition of ox bile and brilliant green to improve selectivity. The addition of novobiocin at 40mg per liter was later described by Jeffries5 to improve inhibition of Proteus species. A selective enrichment medium for the isolation of Salmonella, the formulation conforms to ISO 6579:20025. Presumptive Salmonella should be confirmed using appropriate biochemical and serological techniques. For the complete method, please refer to ISO 6579:20021.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Straw | |
Muller-Kauffmann Tetrathionate-Novobiocin Broth | |
Dehydrated Culture Media | |
Each |
10°C to 30°C | |
Powder | |
500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin Broth (MkTTn) >
Spot an opportunity for improvement?Share a Content Correction