Learn More
Multi 13 elements solution - Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U for ICP 0.001mg/L, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS 003 250
This item is not returnable.
View return policy
Specifications
Thirteen Elements | |
Bottle | |
Limit Detection Multi-component Verification Solution | |
2% HNO3 |
0.001 mg/L | |
250 mL | |
Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U | |
Liquid |