Learn More
Multi 13 elements solution - Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U for ICP 0.001mg/L, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand:  Fisher Chemical MS 003 500
 View more versions of this product
Product Code. 12966183
  • £988.00 / 500mL
Explore more special offers
Specifications
Thirteen Elements
Bottle
Limit Detection Multi-component Verification Solution
2% HNO3
0.001 mg/L
500 mL
Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U