Learn More
Multi 13 elements solution - Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U for ICP 0.001mg/L, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS 012 050
Code : 04
Additional Details : Weight : 0.05000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Specifications
Four Elements (Precious Metals) | |
Bottle | |
ICP/MS Calibration Multi-element Standard Solution | |
10% HCl |
10 mg/L | |
50 mL | |
Ir,Pd,Pt,Ru | |
Liquid |
Safety and Handling
- Multi-element Standard Solution for ICP. 4 Components in HCl 10% (MS 012)
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .