Learn More
Thermo Scientific™ Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Speed up your analysis with high-throughput measurements.
£241.00 - £353.00
Specifications
For Use With (Equipment) | GENESYS 140/150/180, BioMate 160 Spectrophotometers |
---|
Product Code | Brand | Description | For Use With (Equipment) | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Holds | Price | Quantity & Availability | ||||
15871148
|
Thermo Scientific™
840-314700 |
4-position Carousel with Long Path Rectangular Cell Holders. Magnetic latch design for easy carousel exchange. | GENESYS 140/150/180, BioMate 160 Spectrophotometers | Up to 4 Long Pathlength Rectangular Cells (up to 50 mm) |
£353.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856095
|
Thermo Scientific™
840-303600 |
4-position Carousel Base | GENESYS 140/150/180, BioMate 160 Spectrophotometers | Up to 4 10 mm Pathlength Cells |
£241.00
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.Specifications
GENESYS 140/150/180, BioMate 160 Spectrophotometers |