Learn More
Miele Professional™ Mobile Injector Unit E 355
Mobile unit E 355 for the optimum loading of narrow necked glassware
Brand: Miele Professional™ 3809560
1783.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- equipped with 16 injector nozzles
- Half can be equipped with other inserts
Specifications
Multi-purpose Mobile Injector | |
G 7804 and G 7883, Narrow-necked laboratory glassware/flasks/volumetrics | |
16 Jets/clips |
Product Content Correction
Product Title
Miele Professional™ Mobile Injector Unit E 355 > For Use With: G 7804 and G 7883, Narrow-necked laboratory glassware/flasks/volumetrics
Spot an opportunity for improvement?Share a Content Correction