Learn More
ATAGO™ Multi-Wavelength Abbe Refractometer DR-M2
The Multi-Wavelength Abbe Refractometer DR-M2 digitally displays the measurement result of refractive index or Abbe number on the LCD.
Brand:  ATAGO™ 1410
Product Code. 11446248
  • £11575.00 / Each
Explore more special offers
Description
Description
  • Refractive index or Abbe number (νd or νe) can be measured at different wavelengths ranging from 450 to 1100 nm.
  • The DR-M2 digitally displays measurement result of refractive index or Abbe number on the LCD.
  • Measurement can be achieved by matching the boundary line with the intersection point of the cross hairs.
  • This refractometer is connectable with Digital printer DP-63 (B) (Optional).
Specifications
Specifications
Benchtop Refractometer
±0.0002
6.0 kg (Refractometer), 3.2 kg (Light Source Unit)
5°C to 50°C
100/240 V
1.3278 to 1.7379 (at 450 nm), 1.3000 to 1.7100 (at 589 nm), 1.2912 to 1.7011 (at 680 nm), 1.2743 to 1.6840 (at 1100 nm)
LCD
130 x 290 x 310 mm (Refractometer), 150 x 330 x 110 mm (Light Source Unit)
AC100
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
ATAGO™ Multi-Wavelength Abbe Refractometer DR-M2 > Refractometer; Accuracy: ±0.0002