missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ratiolab™ Multibox™ultra Storage System
Click to view available options
Color:
Red
Conditionnement:
Each
Description
- Holds the tips in plastic matrices
- Convenient and safe tip pick-up, even with 8- and 12-channel pipettors
- For tip removal the dust-proof, hinged cover may be opened completely or partially by using the sliding mechanism
- For pipette tips up to 1000 μL
- Sold individually
- Provided 75 to a pack
Spécification
Spécification
| Closure Included | Yes |
| Array | 6 x 10 |
| Autoclavable | Autoclavable |
| Color | Red |
| For Use With (Application) | Storage and sterilization |
| For Use With (Equipment) | Pipette tips up to 1000 μL |
| Quantity | 1 Ea. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu