missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MultiDrive BL 2000 T Blender Vessel

Vessel with mixing knife is particularly useful in combination with IKA MultiDrive basic for wet grinding of a variety of applications, such as meat, sausage, soy beans, silage, vegetables. Integrated temperature sensor allows continuous monitoring and individual setting of the temperature limit.

Brand:  IKA™ 0025003962

Product Code. 16268631

  • £583.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The integrated temperature sensor allows continuous monitoring and an individual setting of the temperature limit. This is especially meaningful for temperature sensitive materials.
  • All parts can be cleaned in a dishwasher. The knife is easy to dismantle for intensive cleaning.
  • The scope of delivery includes a standard mixing knife made of stainless steel.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2000 mL
5 Mohs
Stainless Steel
Max: 7 mm
220 x 180 x 280 mm
Temperature sensor
BL 2000 T
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IKA™ MultiDrive BL 2000 T Blender Vessel > 2000 mL, includes Temperature sensor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.