Learn More
Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Brand: Thermo Scientific™ 24072672
3699.43 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.
Ordering Information:
- Cat. No. 24070297 and 24070290 include tip combs

Specifications
Tube Dispensing Cassette | |
Standard tube dispensing cassette 10-pack | |
5 to 2500 μL | |
<7 mL |
Autoclavable (50 times at 121°C, 1 bar, 20 min.) | |
Polypropylene Tip, Silicone Tubing | |
Multidrop Combi, Multidrop 384, Multidrop DW | |
Tip: 0.5 mm; Tubing: 1.3 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.