Learn More
Thermo Scientific™ Multidrop™ Combi Accessories
Guarantee excellent dispensing results and day-to-day performance with combi accessories.
Brand:  Thermo Scientific™ 24073296
2528.81 GBP valid until 2025-03-14
Use promo code "24150" to get your promotional price.
 View more versions of this product
Product Code. 10095273
  • £2661.91 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi Accessories.
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi Accessories.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube Dispensing Cassette
Small tube metal tip dispensing cassette 5-pack
0.5 to 50 μL
<5 mL
Autoclavable (10 times at 121°C, 1 bar, 20 min.)
Polypropylene Tip, Silicone Tubing
Multidrop Combi, Multidrop Micro
Tip: 0.22; Tubing: 0.4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Multidrop™ Combi Accessories >