Learn More
Thermo Scientific™ Multidrop™ Combi Reagent Dispenser
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.
Brand: Thermo Scientific™ 5840340
18122.00 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.
The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.- Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
- Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
- Visual icon-based graphic display makes it easy to use and program, even without training
- Minimal dead volume and back-flushing reduces reagent costs
- Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
- Full robotic compatibility gives increased throughput
- Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range
Multidrop Combi SMART:
- Adds an advanced built-in tracing system to the Multidrop Combi
- Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability
Recommended for:
- Assay development
- Primary and secondary screening
- Compound storage
- Genomics and proteomics research
- PCR set-up
- Sequencing set-up
- Cell based assays
- Bead based assays
- ELISA assays

Specifications
|
|
Multidrop™ Combi SMART+ Reagent Dispenser | |
The instrument is intended for automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA. | |
One SMART+ standard tube dispensing cassette (cat. no. N22704) | |
6, 12, 24, 48, 96, and 384 well plates | |
|
|
<1mL volume range 0.5-50μL; <7mL volume range >50μL (it is possible to blackflush the reagent into original reservoir to avoid loss of expensive reagent) | |
14 x 8.6 x 12.9 in. (35.5 x 22 x 33 cm) | |
9.1 kg |
Reagent Dispenser | |
8 | |
Can be used with automated systems (e.g. Thermo Scientific™ Orbitor™ Microplate Mover) and 6, 12, 24, 48, 96, or 384-well plate format. It is capable of dispensing into 2D tubes in SLAS format that are shorter than 50mm. | |
On-board software and Serial RS-232 or USB for PC control | |
|
|
|
|
1 year | |
20.1 lb. | |
100 to 240 Vac 50/60Hz |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
836-8C0217SM | Certificate of Analysis | 31/07/2023 | 5840340 |
836-8C0215SM | Certificate of Analysis | 31/07/2023 | 5840340 |
836-8C0208SM | Certificate of Analysis | 31/07/2023 | 5840340 |
836-8C0216SM | Certificate of Analysis | 31/07/2023 | 5840340 |
836-8C0211SM | Certificate of Analysis | 31/07/2023 | 5840340 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.