Learn More
Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Brand: Thermo Scientific™ N22700
864.46 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.
Designed for use with Multidrop Combi SMART+ dispensers Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time Available in standard and small tube versions Distinguished from normal cassettes by RFID tag

Specifications
Autoclavable | |
In contact with liquid: PEEK, silicone, and PP (polypropylene) | |
Automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA. | |
<1 mL |
SMART+ small tube plastic tip dispensing cassette | |
0.5 to 50 μL | |
Multidrop Combi SMART+ instrument with serial numbers 836-8C0001SM onwards |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.