missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Pico 1 & Pico 8 Digital Dispenser
GREENER_CHOICE

Miniaturize assays and achieve accurate results on picolter range with non-contact dispensing with eay to use software

£15915.00 - £55195.00

Specifications

Product Type Digital Dispenser
For Use With (Application) Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
Interface USB
Plate Type 12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
Precision ≤ 8%
View More Specs

Products 2
Product Code Brand Description Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Description Dimensions (L x W x H) Price Quantity & Availability  
16671296
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5840500
Multidrop™ Pico 1 Digital Dispenser 31 x 28 x 20 cm
£15915.00
Each
In Stock
to see stock.
Add to basket
 
16643823
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5840600
Multidrop™ Pico 8 Digital Dispenser 47 x 38 x 23 cm
£55195.00
Each
In Stock
to see stock.
Add to basket
 
Description
Show More
Specifications

Specifications

Digital Dispenser
USB
≤ 8%
100 to 240V 50/60Hz
50/60 Hz
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
11 pL to 10 μL in seconds
100/240 V
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Multidrop™ Pico 1 & Pico 8 Digital Dispenser

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.