Learn More
Invitrogen™ Multiplexed Proteomics™ Phosphoprotein Gel Stain Kits (with Pro-Q™ Diamond and SYPRO™ Ruby Gel Stains)
Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis.
£260.00 - £836.00
Specifications
Detection Location | In-Gel Detection |
---|---|
Detection Method | Fluorescence |
Label or Dye | SYPRO Ruby, Pro-Q Diamond |
Product Type | Phosphoprotein Gel Stain Kit |
Product Line | PRO-Q™, SYPRO™ |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10788273
|
Invitrogen™
M33305 |
Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit #1 |
£836.00
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10104722
|
Invitrogen™
M33306 |
Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit #2 |
£260.00
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis.
Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis. This convenient Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Protein Gel Stain Pack provides each stain in 1-liter amounts (Cat. No. M-33305) or in 200-mL amounts (Cat. No. M-33306).Features include:
• Sensitive—Pro-Q Diamond Phosphoprotein Gel Stain offers detection level down to 1–16 ng of phosphoprotein per band
• Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. SYPRO Ruby Gel Stain is subsequently used for total protein staining and simplifies the determination of phosphorylation level
• Linear signal—quantify with ease as Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Gel Stain exhibit excellent linearity over three orders of magnitude, ideal for quantitation
Specifications
In-Gel Detection | |
SYPRO Ruby, Pro-Q Diamond | |
PRO-Q™, SYPRO™ | |
Proteins (Phosphoproteins) |
Fluorescence | |
Phosphoprotein Gel Stain Kit | |
Room Temperature |