missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Marigold™ Multiplus 40 Gloves
Click to view available options
Size:
8
9
10
11
Description
- Fully coated in high performance PVC/nitrile
- 100% cotton interlock liner on palm, thumb and forefinger
- Resistance to abrasion and tearing
- Superior plasticized plastoprene integral base coating, made of mix of resins and plasticizers with incorporated elastomers
- Anatomical shape for increased dexterity and comfort
- Ieal for tasks involving mechanical and chemical hazards: maintenance, cleaning, petrochemical plants, factories.
Specifications
Description | Not Anti-static, Not silicon-free |
Coating Material | PVC |
Color | Blue |
Latex Free | No |
Material | PVC |
Product Type | Industrial Gloves |
Size | 10 |
Glove Cuff Style | Gauntlet |
Liner Material | Interlock cotton |
Lining | Color: Blue |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'