missing translation for 'onlineSavingsMsg'
Learn More
Multipurpose alkaline detergent, RBS 35 concentrate, J.T. Baker™ Product Code.: 10002332
compliance-icons

Multipurpose alkaline detergent, RBS 35 concentrate, J.T. Baker™

Product Code. 10002332
5 L
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy

Product Code. 10002332

Brand: JT Baker 7348.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for manual cleaning, soaking and ultrasonic baths. These biodegradable surfactants are alkaline, foaming and insensitive to water hardness.

  • High wetting and emulsifying power
  • No risk of calcareous deposits when diluted in tap water
  • Biodegradable
  • Total rinse
  • Suitable for cleaning glassware, laboratory equipments and for cleaning parts made of glass, stainless steel, injection moulds and production equipment.
  • Typical concentration for use: 2%

Specifications

pH 11.8 (2%)
Quantity 5 L
Packaging Polyethyelene bottle
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Multipurpose alkaline detergent, RBS 35 concentrate, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.