Learn More
Syngene™ Multipurpose Bonded Layered Carbon Filter
The Multipurpose Bonded Layered Carbon Filter is used to absorb fumes and odors.
Brand:  Syngene™ SYN-ACF431
Additional Details : Weight : 3.62870kg
 View more versions of this product
Product Code. 17725964
  • £1215.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Description
Description
AirClean™ Systems manufactures gas phase bonded carbon and multi-layered filters that are available with standard coconut shell carbon or chemically impregnated carbon, depending on your application. AirClean™ Systems gas-phase, coconut shell carbon filters utilize our unique bonded carbon filtration media. This minimizes the release of potentially hazardous carbon dust found in traditional granular carbon filters. Our proprietary bonding process holds carbon in a predictable matrix, preventing carbon shift that can lead to 'dead spots', common in traditional granular filters.
Features
  • Bonded carbon — no filter dusting
  • Even distribution of impregnates, if applicable
  • Multi-layering options for larger application compatibility
Specifications
Specifications
Carbon
3 in.
1 Ea.
30.5 cm
12 in.
24 in.
8 cm
61 cm