Learn More
Axygen™ MultiRack Empty Rack
MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.
£100.62 - £102.00
Specifications
Certifications/Compliance ISO 13845
Autoclavable Autoclavable
Color Clear
Material Polyethylene
Sterility Non-sterile
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
16372237
View Documents
Axygen™
MR-R-A
10 μL XT, 200 μL, 200 μL NX, 300 μL
£102.00
Pack of 5
Estimated Shipment: 16-09-2025
to see stock.
Add to basket
16382237
View Documents
Axygen™
MR-R-B
200 μL XT, 200 μL XT, 300 μL XT, 1000 μL XT
£100.62
Pack of 5
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Description
Description
  • Detectable DNase/RNase/Human DNA/PCR inhibitor free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Specifications
Specifications
ISO 13845
Clear
Non-sterile
5/Pk.
Autoclavable
Polyethylene
1/Cs.
5/pk, 1 pk/cs
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Axygen™ MultiRack Empty Rack