missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ MultiRack Reload Tip System

Product Code. 16302227 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1250 μL
Tip Style:
Extended Length
Universal
Rack Color:
Blue
Green
Red
Yellow
Unit Size:
Pack of 768
Pack of 960
This item is not returnable. View return policy

Product Code. 16302227

Brand: Axygen™ MR10XTRL

Only null left
Add to basket
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.

The Axygen MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter, and reload packaging styles to meet the variety of needs in the lab setting. The MultiRack tip system offers standard and extended length tips covering volumes from 1 μL to 1250 μL. For racked and reload packaging styles, the tips are pre-loaded into easy-load color-coded deck inserts. With the smaller footprint and reusable rack, this system uses 30% less plastic, minimizing the environmental impact. The stackable design saves critical bench space. Axygen MultiRack Tip System products are made in the USA and meet ISO standards. All tips are free of detectable RNase, DNase, DNA, and pyrogens. For more details visit our Certificates page at www.corning.com/lifesciences.
Features
• Reusable rack with reload systems 
• Smaller footprint and stackable design, saving valuable bench space
• Extended tips offered in 10, 300, and 1250 μL (1000 μL extended length) volumes
• Color-coded racks for easy volume identification

 

TRUSTED_SUSTAINABILITY

Especificaciones

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Red
Rack Color Red
Autoclavable Autoclavable
Certifications/Compliance ISO 13845
Clean Claims Free of detectable DNAse/RNAse, human DNA, and PCR inhibition
Format Refill
Tip Style Extended Length
Packaging Reload System
Quantity 960/pk, 4 pks/cs
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line MultiRack
Material Polyethylene
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado