missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS Durapore™ 96-Well Filter Plates
Designed for full automation compatibility. MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.
£361.00 - £1970.00
Specifications
Certifications/Compliance | ANSI, SBS |
---|---|
Format | Standard |
Membrane | Hydrophilic PVDF |
Array | 8 x 12 |
No. of Wells | 96 |
Product Code | Brand | Color | Material | Quantity | Pore Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Quantity | Pore Size | Price | Quantity & Availability | ||
10011953
|
Merck
MSBVN1B50 |
Opaque | Barex | 50/Pk. | 1.2 μm |
£1715.00
Pack of 50 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||
10659113
|
Merck
MSBVN1250 |
Clear | Styrene | 50/Pk. | 1.2 μm |
£1790.00
Pack of 50 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
10777264
|
Merck
MSBVS1210 |
Opaque | Styrene | 10/Pk. | 1.2 μm |
£525.00
Pack of 10 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
10432753
|
Merck
MSBVN1210 |
Clear | Styrene | 10/Pk. | 1.2 μm |
£601.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
10400334
|
Merck
MSDVN6B50 |
Opaque | Barex | 50/Pk. | 0.65 μm |
£1955.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
10367123
|
Merck
MSGVN2210 |
Clear | Styrene | 10/Pk. | 0.22 μm |
£361.00
Pack of 10 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
||
10154213
|
Merck
MSGVN2250 |
Clear | Styrene | 50/Pk. | 0.22 μm |
£1790.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
10347133
|
Merck
MSGVN2B50 |
Opaque | Barex | 50/Pk. | 0.22 μm |
£1955.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
10715164
|
Merck
MSHVN4B10 |
Opaque | Barex | 10/Pk. | 0.45 μm |
£460.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
10367513
|
Merck
MSHVS4510 |
Clear | Styrene | 10/Pk. | 0.45 μm |
£753.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
10408993
|
Merck
MSHVN4B50 |
Clear | Barex | 50/Pk. | 0.45 μm |
£1970.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
Description
Designed for full automation compatibility. MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.
For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.- Standardized dimensions and aligned, rigid sidewalls
- Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
- Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
- Extended skirt and vacuum manifolds
- Allows easy, contamination-free stacking directly with collection plates
- Protects recessed underdrain
- Provides ample surface for bar code labels
- Improves vacuum seal
- Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
- Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
- Opaque plastic prevents light transfer between wells during direct scintillation counting
- Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
- Meet ANSI™/SBS 2004 plate specifications
Specifications
ANSI, SBS | |
Hydrophilic PVDF | |
96 |
Standard | |
8 x 12 |