Learn More
Thermo Scientific™ Multiskan™ FC Microplate Photometer
Multiskan FC microplate photometer is a reliable instrument for a wide variety of research and routine applications. It reads 96- and 384-well plates and is equipped with shaking and incubation capabilities for temperature critical assays. It can be used as a stand-alone instrument or under PC control with intuitive Thermo Scientific SkanIt™ software.
£6040.00 - £8360.00
Product Code | Brand | Description | Plate Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Plate Type | Price | Quantity & Availability | ||||
11590685
|
Thermo Scientific™
51119000 |
Multiskan FC | 96-well Plate |
£6040.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11500695
|
Thermo Scientific™
51119100 |
Multiskan FC with incubator | 96 and 384-well Plates |
£8360.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Multiskan FC brings together over 40 years experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.
- Proven performance and reliable day-to-day results through patented optical design and in-built self-diagnostics
- A broad wavelength range of 340–850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
- Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm, preinstalled
- A comprehensive range of easy-to install additional filters available for order
- Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
- Shaking and incubation up to 50°C for temperature critical assays
- Ease of use through the large color screen and a variety of language versions
- Visual and logical SkanIt software for comprehensive instrument control and data handling
- Multiskan verification plate available for instrument performance verification
- Robot compatibility for high-throughput environments
Warranty
1 Year
Specifications
Multiskan |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.