Learn More
Fisherbrand™ MultiSUB Midi 96 Horizontal Gel System
Fisherbrand™ MultiSUB Midi 96 Horizontal Gel System, 96-well comb block and specialised tray replicates the format and well-spacing of microplates and thermal cycler blocks.
Brand:  Fisherbrand™ MSMIDI96ST/2M
Product Code. 12104011
  • £395.00 / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for loading of DNA samples from multi-well plates
  • Ideal for high throughput electrophoresis of PCR products or samples containing a small number of DNA bands
  • Average run-time is just 15 to 30 minutes
  • Direct microplate format for easy lane identification
  • Multichannel pipette compatible combs for speed loading
  • Stretch versions available with extended 3.6cm separation distance to double capacity
Specifications
Specifications
Horizontal Gel Electrophoresis System
Multi Sub Midi96 STRETCH, UV tray, Comb block with 12 x 8 sample, 1mm thick combs, casting dams
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ MultiSUB Midi 96 Horizontal Gel System > MultiSUB Midi 96 Gel System