missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ multiSUB™ Midi Horizontal Gel System Product Code.: 12307287
Only undefined left
Add to basket

Fisherbrand™ multiSUB™ Midi Horizontal Gel System

Product Code. p-7083382 Shop All Fisherbrand Products
50, W x L: 10 x 7 cm, Each
Click to view available options
Sample Size:
50
100
Gel Dimensions:
W x L: 10 x 10 cm
W x L: 10 x 7 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12307287

Only undefined left
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy

Designed for routine horizontal gel electrophoresis.

  • Suitable for rapid electrophoresis
  • Low buffer volumes
  • Runs about 50 to 100 samples

Specifications

Type Horizontal Gel Electrophoresis System
Gel Dimensions W x L: 10 x 7 cm
Includes multiSUB™ Midi electrophoresis unit, 10 x 7cm UV Tray, 2 x 16 sample combs, loading guides and dams
Sample Size 50
Volume (English) Buffer 0.07 gal.
Volume (Metric) Buffer 300 mL
Length (English) 4.92 in.
Length (Metric) 12.5 cm
Width (English) 8.66 in.
Width (Metric) 22 cm
Height (English) 3.54 in.
Height (Metric) 9 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ multiSUB™ Midi Horizontal Gel System >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.