Description
- Runs up to 100 samples
- Low buffer volumes
- Injection-moulded construction
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis Unit | |
100 | |
220 x 125 x 90 mm |
70L x 100 mm W, 100L x 100 mm W | |
300 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
-
Fisherbrand Meet the Family
Product Content Correction
Product Title
Fisherbrand™ Sub-Gel Midi Horizontal Gel Electrophoresis Unit >
Spot an opportunity for improvement?Share a Content Correction