missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ multiSUB™ Midi96 STRETCH Horizontal Gel System
Suitable for high throughput electrophoresis of PCR products or samples containing small number of DNA bands.
£381.00
Specifications
Running Time | 15 to 30 min. |
---|---|
Type | Horizontal Gel Electrophoresis System |
Gel Dimensions | W x L: 10 x 24 cm (3.93 x 9.44 in.) |
Sample Size | 96 |
Volume (English) Buffer | 0.18 gal. |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12327287
|
Fisherbrand™
|
multiSUB™ Midi96 STRETCH electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with two marker lanes, casting dams |
£381.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
12337287
|
Fisherbrand™
|
multiSUB™ Midi96 STRETCH electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with one marker lane, casting dams |
£381.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
Description
- 96-well comb block format
- Standard microplate configuration with additional lanes for markers Offset layout of wells allows maximum run length of 1.8cm per well which results in improved separation
- Simplified leak-proof casting dams
- Ultra-compact size minimizes buffer usage and bench space requirements
Specifications
15 to 30 min. | |
W x L: 10 x 24 cm (3.93 x 9.44 in.) | |
0.18 gal. | |
18.30 in. | |
3.14 in. |
Horizontal Gel Electrophoresis System | |
96 | |
700 mL | |
46.5 cm | |
8 cm |