Learn More
BD Biosciences™ Multitest CD3/CD16+CD56/CD45/CD19 (CE/IVD)
Four-color direct immunofluorescence reagent for identifying and determining the percentages of human T, natural killer, and B lymphyocytes in erhythrocyte-lysed whole blood.
Brand: BD Biosciences™ 342416
Additional Details : Weight : 0.05710kg
Description
- Four-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T (CD3+), natural killer (NK) (CD3–CD16+CD56+), and B (CD3–CD19+) lymphocytes in erythrocyte-lysed whole blood.
- When used with BD Trucount™ tubes, absolute counts of these populations can be enumerated from a single tube.
- BD Multitest CD3/CD16+CD56/CD45/CD19 and BD Trucount tubes can be used with the BD FACS™ Loader, BD FACSVia™ Loader, and BD FACS™ Universal Loader.
Specifications
Trucount™ Tubes | |
50 Tests | |
CE/IVD |
Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. | |
Immunofluorescence Flow Cytometry Test Reagents |