missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide, For Complexometry, Honeywell Fluka™
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | >300°C |
UN Number | NONH for all modes of transport |
Beilstein | 3831445 |
Formula Weight | 284.19g/mol |
Packaging | Glass Bottle |
Color | Red |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Murexide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'