missing translation for 'onlineSavingsMsg'
Learn More

Muscimol, Tocris Bioscience™

Product Code. 30001383
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent GABAA agonist; also GABAA-ρ partial agonist

Potent GABAA receptor agonist and partial GABAA-ρ receptor agonist. Inhibits memory retention and attenuates airway constriction in vivo.

Chemical Identifiers

CAS 2763-96-4
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00057894
InChI Key ZJQHPWUVQPJPQT-UHFFFAOYSA-N
Synonym muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole
PubChem CID 4266
ChEBI CHEBI:7035
IUPAC Name 5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one
SMILES NCC1=CC(=O)NO1

Specifications

Quantity 1 mg
Formula Weight 114.1
Percent Purity >98%
Chemical Name or Material Muscimol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'