Learn More
Elma™ Mutebox
Product Code. 17841727
Click to view available options
:
Mutebox; For Use With (Equipment): Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Med, Elmasonic Select, Elmasonic EASY, Elmasonic xtra TT
Product Code. 17841727
Brand: Elma™ 1111901
Only null left
Add to basket
Add to basket
Reduce noise levels by up to 85% with the Mutebox.
  • Guarantees low-noise cleaning
  • Quiet fan and optimal insulation make working with ultrasonic equipment a pleasant, health-friendly and quiet experience, whether for cleaning or sample preparation
  • Fan extracts condensation water and vapors to the rear
  • Interior lighting and glass door let you keep an eye on the cleaning process at all times
  • Two independent access points ensure that the items being cleaned and the ultrasonic unit are always easily accessible
  • Made of high-quality steel and is easy to clean, ensuring hygienic operation
Specifications
Type Mutebox
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Med, Elmasonic Select, Elmasonic EASY, Elmasonic xtra TT
Material Stainless Steel