Learn More
Honeywell™ Mutex E Light Protective Clothing
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Antistatic
- Silicone and latex free
- Excellent ergonomics and fitting
- 3-piece hood
- Special crotch design
- Outside flap and dual slider sipper
- Breathability
- 3-layer polypropylene
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Chemical-resistant Coveralls |
Size | 3X-Large |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Protection Class | Protection type: 5, 6 |
Thickness | 56 gsm |